Isoamyl gallate ≥98.0%

Supplier: TCI America

Synonyms: Gallic Acid Isoamyl Ester

G0012-500G G0012-25G
TCG0012-500GEA 1025.42 CAD
TCG0012-500G TCG0012-025G
Isoamyl gallate ≥98.0%
Isoamyl gallate

CAS Number: 2486-02-4
MDL Number: MFCD00016431
Molecular Formula: C12H16O5
Molecular Weight: 240.26
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 147


Other Product Names / Synonyms: Gallic Acid Isoamyl Ester

Formula: C₁₂H₁₆O₅
MW: 240.26 g/mol
Melting Pt: 147 °C
MDL Number: MFCD00016431
CAS Number: 2486-02-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4373fbe6b884c367a889583390ec4469d6c7a1ea&vid=4373fbe6b884c367a889583390ec4469d6c7a1ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR