8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid ≥97.0%

Supplier: TCI America

Synonyms: Fmoc-NH-PEG2-CH2COOH, 8-(Fmoc-amino)-3,6-dioxa-n-octanoic acid, Fmoc-8-amino-3,6-dioxaoctanoic acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic acid, 1-(9H-Fluoren-9-yl)-3-oxo-2,7,10-trioxa-4-azadodecan-12-oic acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic acid

F0719-1G F0719-200MG
TCF0719-1GEA 420.28 CAD
TCF0719-1G TCF0719-200MG
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid ≥97.0%
8-[(9H-Fluoren-9-ylmethoxy)carbonylamino]-3,6-dioxa-n-octanoic acid

CAS Number: 166108-71-0
MDL Number: MFCD01321015
Molecular Formula: C21H23NO6
Molecular Weight: 385.42
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Crosslinkers, PEGylation Reagents & PEG Spacers
Other Product Names / Synonyms: 8-(Fmoc-amino)-3,6-dioxa-n-octanoic Acid, [2-[2-[(9H-Fluoren-9-ylmethoxy)carbonylamino]ethoxy]ethoxy]acetic Acid, [2-[2-[(Fmoc-amino)ethoxy]ethoxy]acetic Acid, Fmoc-NH-PEG2-CH2COOH

Formula: C₂₁H₂₃NO₆
MW: 385.42 g/mol
MDL Number: MFCD01321015
CAS Number: 166108-71-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afee46233aa0097a23700426283b8bcc83621780&vid=afee46233aa0097a23700426283b8bcc83621780&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR