1-(2-Furoyl)piperazine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Furan-2-yl(piperazin-1-yl)methanone hydrochloride

F0705-25G F0705-5G
TCF0705-25GEA 123.24 CAD
TCF0705-25G TCF0705-5G
1-(2-Furoyl)piperazine hydrochloride ≥98.0%
1-(2-Furoyl)piperazine hydrochloride

CAS Number: 60548-09-6
MDL Number: MFCD00236012
Molecular Formula: C9H12N2O2
Molecular Weight: 216.67
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 207


Other Product Names / Synonyms: 1-[(2-Furan)carbonyl]piperazine Hydrochloride

Formula: C₉H₁₃ClN₂O₂
MW: 216.6647 g/mol
Melting Pt: 207 °C
MDL Number: MFCD00236012
CAS Number: 60548-09-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa0c04857ea086654ec1be03ba961dc3f7004f62&vid=fa0c04857ea086654ec1be03ba961dc3f7004f62&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR