9-Fluorenylmethyl carbamate ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America

F0689-5G F0689-1G
TCF0689-5GEA 225.94 CAD
TCF0689-5G TCF0689-1G
9-Fluorenylmethyl carbamate ≥98.0% (by HPLC, total nitrogen)
9-Fluorenylmethyl carbamate

CAS Number: 84418-43-9
MDL Number: MFCD00270532
Molecular Formula: C15H13NO2
Molecular Weight: 239.27
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 204


Product Categories: Synthetic Organic Chemistry, Protection & Derivatization Reagents (for Synthesis)
Other Product Names / Synonyms: Carbamic Acid 9-Fluorenylmethyl Ester, (9H-Fluoren-9-ylmethoxy)carboxamide, Fmoc-NH2

Formula: C₁₅H₁₃NO₂
Melting Pt: 204 °C
MDL Number: MFCD00237376
CAS Number: 84418-43-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c0bccebe675bd97a73c317c1ba62aec39eae35d&vid=5c0bccebe675bd97a73c317c1ba62aec39eae35d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR