2-Fluoro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
Synonyms:
2-Fluoro-4-methylbenzeneboronic acid, (2-Fluoro-4-methylphenyl)boronic acid
F0683-5G
F0683-1G
TCF0683-5GEA
105.86
CAD
TCF0683-5G
TCF0683-1G
2-Fluoro-4-methylphenylboronic acid (contains varying amounts of Anhydride)
2-Fluoro-4-methylphenylboronic acid
CAS Number: 170981-26-7
MDL Number: MFCD05664239
Molecular Formula: C7H8BFO2
Molecular Weight: 153.95
Form: Crystal
Color: White
Melting point (°C): 230
Other Product Names / Synonyms: 2-Fluoro-4-methylbenzeneboronic Acid
Formula:
C₇H₈BFO₂ MW: 153.95 g/mol Melting Pt: 230 °C |
MDL Number:
MFCD05664239 CAS Number: 170981-26-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...