N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-2-phenylglycine ≥98.0%
Supplier: TCI America
Synonyms:
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-2-phenylacetic acid, N-Fmoc-L-α-phenylglycine, Fmoc-Phg-OH, N-Fmoc-L-2-phenylglycine
CAS Number: 102410-65-1
MDL Number: MFCD00155632
Molecular Formula: C23H19NO4
Molecular Weight: 373.41
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 176
Specific rotation [a]20/D: 81 deg (C=1, DMF)
Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-2-phenylglycine, Fmoc-Phg-OH
Formula:
C₂₃H₁₉NO₄ Melting Pt: 176 °C |
MDL Number:
MFCD00155632 CAS Number: 102410-65-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...