Flutamide ≥98.0%

Supplier: TCI America

F0663-25G F0663-5G
TCF0663-25GEA 213.3 CAD
TCF0663-25G TCF0663-5G
Flutamide ≥98.0%
Flutamide

CAS Number: 13311-84-7
MDL Number: MFCD00072009
Molecular Formula: C11H11F3N2O3
Molecular Weight: 276.22
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 112


Product Categories: Synthetic Organic Chemistry, Fluorinated Bioactive Compounds, Bioscience, Antitumor Ingredients
Other Product Names / Synonyms: 4'-Nitro-3'-(trifluoromethyl)isobutyranilide, 2-Methyl-N-[4-nitro-3-(trifluoromethyl)phenyl]propionamide

Formula: C₁₁H₁₁F₃N₂O₃
Melting Pt: 112 °C
MDL Number: MFCD00072009
CAS Number: 13311-84-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a2b28b5a252f25171df4244b5d915ae9696e0184&vid=a2b28b5a252f25171df4244b5d915ae9696e0184&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR