2-Fluoro-3-nitrotoluene ≥98.0%
Supplier: TCI America
F0661-25G
F0661-5G
TCF0661-25GEA
206.98
CAD
TCF0661-25G
TCF0661-5G
2-Fluoro-3-nitrotoluene ≥98.0%
2-Fluoro-3-nitrotoluene
CAS Number: 437-86-5
MDL Number: MFCD03412242
Molecular Formula: C7H6FNO2
Molecular Weight: 155.13
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 111
Melting point (°C): 19
Specific Gravity (20/20): 1.28
Formula:
C₇H₆FNO₂ Boiling Pt: 111 °C Melting Pt: 19 °C Density: 1.28 (20 °C) |
MDL Number:
MFCD03412242 CAS Number: 437-86-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...