(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid ≥98.0%

Supplier: TCI America

Synonyms: N-Fmoc-L-citrulline, Nalpha-Fmoc-L-citrulline, Fmoc-Cit-OH

F0626-5G F0626-1G
TCF0626-5GEA 114.08 CAD
TCF0626-5G TCF0626-1G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid ≥98.0%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-ureidopentanoic acid

CAS Number: 133174-15-9
MDL Number: MFCD00151943
Molecular Formula: C21H23N3O5
Molecular Weight: 397.43
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -10 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-citrulline

Formula: C₂₁H₂₃N₃O₅
MDL Number: MFCD00151943
CAS Number: 133174-15-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=49b326a691f43d06d1c1edddb3c952f2e0706e88&vid=49b326a691f43d06d1c1edddb3c952f2e0706e88&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR