N-Fmoc-D-valine ≥98.0%

Supplier: TCI America

Synonyms: Fmoc-D-Val-OH, D-Valine, N-FMOC protected

F0610-25G F0610-5G
TCF0610-25GEA 1055.44 CAD
TCF0610-25G TCF0610-5G
N-Fmoc-D-valine ≥98.0%
N-Fmoc-D-valine

CAS Number: 84624-17-9
MDL Number: MFCD00062953
Molecular Formula: C20H21NO4
Molecular Weight: 339.39
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 146
Specific rotation [a]20/D: 17 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-valine, Fmoc-D-Val-OH

Formula: C₂₀H₂₁NO₄
MW: 339.39 g/mol
Melting Pt: 146 °C
MDL Number: MFCD00062953
CAS Number: 84624-17-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f4a411a26024b9dc3409a360ea7677e229f15f0a&vid=f4a411a26024b9dc3409a360ea7677e229f15f0a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR