N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine ≥98.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
(2R,3S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoic acid, N-Fmoc-D-threonine, Fmoc-D-Thr-OH
F0608-1G
F0608-5G
TCF0608-1GEA
75.84
CAD
TCF0608-1G
TCF0608-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-threonine
CAS Number: 157355-81-2
Molecular Formula: C19H19NO5
Molecular Weight: 341.36
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 15 deg (C=1, DMF)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-threonine, Fmoc-D-Thr-OH
Formula:
C₁₉H₁₉NO₅ MW: 341.36 g/mol |
MDL Number:
MFCD00153365 CAS Number: 157355-81-2 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...