4-Aminononafluorobiphenyl ≥96.0%

Supplier: TCI America

A1346-5G A1346-1G
TCA1346-005GEA 646.22 CAD
TCA1346-005G TCA1346-1G
4-Aminononafluorobiphenyl ≥96.0%
4-Aminononafluorobiphenyl

CAS Number: 969-25-5
MDL Number: MFCD00671483
Molecular Formula: C12H2F9N
Molecular Weight: 331.14
Purity/Analysis Method: >96.0% (GC)
Form: Crystal
Melting point (°C): 146

Formula: C₁₂H₂F₉N
Melting Pt: 146 °C
MDL Number: MFCD00671483
CAS Number: 969-25-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=660c06d6bddef704946d3530b47dd359e662f85b&vid=660c06d6bddef704946d3530b47dd359e662f85b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR