N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

Synonyms: N-Fmoc-D-methionine, Fmoc-D-Met-OH

F0594-5G F0594-1G
TCF0594-5GEA 84.37 CAD
TCF0594-5G TCF0594-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine ≥98.0% (by HPLC, titration analysis)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-methionine

CAS Number: 112883-40-6
MDL Number: MFCD00062958
Molecular Formula: C20H21NO4S
Molecular Weight: 371.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 132
Specific rotation [a]20/D: 30 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-methionine, Fmoc-D-Met-OH

Formula: C₂₀H₂₁NO₄S
MW: 371.45 g/mol
Melting Pt: 132 °C
MDL Number: MFCD00062958
CAS Number: 112883-40-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff67d8512df2abe684ce2fcc7d357b5657fb75e3&vid=ff67d8512df2abe684ce2fcc7d357b5657fb75e3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR