N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid ≥98.0%
Supplier: TCI America
Synonyms:
Fmoc-D-Asp-OH, N-Fmoc-D-aspartic Acid
F0592-1G
F0592-5G
TCF0592-1GEA
42.66
CAD
TCF0592-1G
TCF0592-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid ≥98.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-aspartic acid
CAS Number: 136083-57-3
MDL Number: MFCD01318740
Molecular Formula: C19H17NO6
Molecular Weight: 355.35
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: 29 deg (C=1, DMF)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-D-aspartic Acid, Fmoc-D-Asp-OH
Formula:
C₁₉H₁₇NO₆ MW: 355.35 g/mol |
MDL Number:
MFCD01318740 CAS Number: 136083-57-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...