Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America

F0586-1G F0586-5G
TCF0586-1GEA 63.2 CAD
TCF0586-1G TCF0586-5G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride ≥98.0% (by HPLC)
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-lysine hydrochloride

CAS Number: 139262-23-0
MDL Number: MFCD00190889
Molecular Formula: C21H24N2O4
Molecular Weight: 404.89
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -11 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-lysine Hydrochloride, Fmoc-Lys-OH.HCl

Formula: C₂₁H₂₄N₂O₄
MDL Number: MFCD00190889
CAS Number: 139262-23-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1744092f99647f16c750b0d7b38de29981c3f620&vid=1744092f99647f16c750b0d7b38de29981c3f620&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR