2-Fluoro-4-hydroxybenzoic acid ≥98.0%

Supplier: TCI America

F0571-5G F0571-25G
TCF0571-5GEA 71.1 CAD
TCF0571-5G TCF0571-25G
2-Fluoro-4-hydroxybenzoic acid ≥98.0%
2-Fluoro-4-hydroxybenzoic acid

CAS Number: 65145-13-3
MDL Number: MFCD01310985
Molecular Formula: C7H5FO3
Molecular Weight: 156.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 202

Formula: C₇H₅FO₃
Melting Pt: 202 °C
MDL Number: MFCD01310985
CAS Number: 65145-13-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4bd246408a5c36d3f3f10046a6f35361ccbebaa&vid=e4bd246408a5c36d3f3f10046a6f35361ccbebaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR