4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥97.0% free radical

Supplier: TCI America

A1343-1G A1343-5G
TCA1343-001GEA 147.9 CAD
TCA1343-001G TCA1343-005G
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-amino-TEMPO) ≥97.0% free radical
4-Amino-2,2,6,6-tetramethylpiperidinooxy (4-Amino-TEMPO, free radical)

CAS Number: 14691-88-4
MDL Number: MFCD00006479
Molecular Formula: C9H19N2O
Molecular Weight: 171.26
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 98
Freezing point (°C): 39
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Organic Radicals (Magnetic Materials), Precise Polymerization Reagents, Analytical Science, Spin Labels
Other Product Names / Synonyms: 4-Amino-TEMPO

Formula: C₉H₁₉N₂O
Boiling Pt: 98 °C
MDL Number: MFCD00006479
CAS Number: 14691-88-4
EINECS: 238-738-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b3b99ab50e7cd6146706d8e848a4ed4aa34d9fbf&vid=26a8e786d68ae1b52b43c79fbec8d1da7c03fb76&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR