4-Fluoro-3-phenoxybenzaldehyde ≥97.0%

Supplier: TCI America

F0564-5G F0564-25G
TCF0564-5GEA 238.58 CAD
TCF0564-5G TCF0564-25G
4-Fluoro-3-phenoxybenzaldehyde ≥97.0%
4-Fluoro-3-phenoxybenzaldehyde

CAS Number: 68359-57-9
MDL Number: MFCD01318148
Molecular Formula: C13H9FO2
Molecular Weight: 216.21
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 135
Flash Point (°C): 110
Specific Gravity (20/20): 1.23


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 2-Fluoro-5-formyldiphenyl Ether

Formula: C₁₃H₉FO₂
MW: 216.21 g/mol
Boiling Pt: 135 °C
Density: 1.23 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD01318148
CAS Number: 68359-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=828688162c49f324316ef79547a5f4e148fd2f45&vid=828688162c49f324316ef79547a5f4e148fd2f45&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR