5-Fluoro-2-methoxy-4-pyrimidinone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0547-25G F0547-5G
TCF0547-25GEA 84.37 CAD
TCF0547-25G TCF0547-5G
5-Fluoro-2-methoxy-4-pyrimidinone ≥98.0% (by GC, titration analysis)
5-Fluoro-2-methoxy-4-pyrimidinone

CAS Number: 1480-96-2
Molecular Formula: C5H5FN2O2
Molecular Weight: 144.11
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 200

Formula: C₅H₅FN₂O₂
Melting Pt: 200 °C
MDL Number: MFCD03265437
CAS Number: 1480-96-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=492b61488c9c024dd15ab41af2031b3f8bf6c0bb&vid=492b61488c9c024dd15ab41af2031b3f8bf6c0bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR