4-(4-Fluorophenoxy)benzonitrile ≥98.0%
Supplier: TCI America
F0532-1G
F0532-5G
TCF0532-1GEA
36.34
CAD
TCF0532-1G
TCF0532-5G
4-(4-Fluorophenoxy)benzonitrile ≥98.0%
4-(4-Fluorophenoxy)benzonitrile
CAS Number: 215589-24-5
Molecular Formula: C13H8FNO
Molecular Weight: 213.21
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 69
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 4-Cyano-4'-fluorodiphenyl Ether
Formula:
C₁₃H₈FNO |
MDL Number:
MFCD01815090 CAS Number: 215589-24-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...