7-Fluoroquinaldine ≥98.0% (by titrimetric analysis)
Supplier: TCI America
F0519-5G
F0519-1G
TCF0519-5GEA
619.83
CAD
TCF0519-5G
TCF0519-1G
7-Fluoroquinaldine ≥98.0% (by titrimetric analysis)
7-Fluoroquinaldine
CAS Number: 1128-74-1
Molecular Formula: C10H8FN
Molecular Weight: 161.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Boiling point (°C): 120
Melting point (°C): 47
Flash Point (°C): 91
Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 7-Fluoroquinaldine
Formula:
C₁₀H₈FN Boiling Pt: 120 °C Melting Pt: 47 °C Flash Pt: 91 °C |
MDL Number:
MFCD02683029 CAS Number: 1128-74-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...