(1-Fluorovinyl)methyldiphenylsilane ≥96.0%

Supplier: TCI America

Synonyms: 1-Fluoro-1-(methyldiphenylsilyl)ethylene

F0518-1G F0518-5G
TCF0518-1GEA 294.1 CAD
TCF0518-1G TCF0518-5G
(1-Fluorovinyl)methyldiphenylsilane ≥96.0%
(1-Fluorovinyl)methyldiphenylsilane

CAS Number: 257610-49-4
Molecular Formula: C15H15FSi
Molecular Weight: 242.37
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.06


Product Categories: Synthetic Organic Chemistry, Vinylation, Silicon Compounds (for Synthesis)
Other Product Names / Synonyms: 1-Fluoro-1-(methyldiphenylsilyl)ethylene

Formula: C₁₅H₁₅FSi
MW: 242.37 g/mol
Density: 1.06 (20 °C)
CAS Number: 257610-49-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=51c27ecabfd9ee4fb1c5bf0dbe0daba6b8b20632&vid=51c27ecabfd9ee4fb1c5bf0dbe0daba6b8b20632&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 96.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR