Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine-tert-butyl ester ≥98.0%

Supplier: TCI America

Synonyms: Nɑ-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu, (2S,3R)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxybutanoate, Nα-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine tert-Butyl Ester

F0517-5G F0517-1G
TCF0517-5GEA 1252.94 CAD
TCF0517-5G TCF0517-1G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine-tert-butyl ester ≥98.0%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-threonine-tert-butyl ester

CAS Number: 120791-76-6
Molecular Formula: C23H27NO5
Molecular Weight: 397.47
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 100
Specific rotation [a]20/D: -4 deg (C=1, CHCl3)
Storage Temperature: 0-10°C


Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-threonine tert-Butyl Ester, Fmoc-Thr-OtBu

Formula: C₂₃H₂₇NO₅
MW: 397.47 g/mol
Melting Pt: 100 °C
MDL Number: MFCD08276378
CAS Number: 120791-76-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6d55210826b90fc00abea55ba4a875ff1fc40746&vid=6d55210826b90fc00abea55ba4a875ff1fc40746&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR