Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-serine-tert-butyl ester ≥98.0%
Supplier: TCI America
Synonyms:
Fmoc-Ser-OtBu, (S)-tert-Butyl 2-((((9H-fluoren-9-yl)methoxy)carbonyl)amino)-3-hydroxypropanoate, Nɑ-Fmoc-L-serine tert-Butyl Ester
CAS Number: 110797-35-8
Molecular Formula: C22H25NO5
Molecular Weight: 383.44
Purity/Analysis Method: >98.0% (HPLC,N)
Form: Crystal
Melting point (°C): 127
Specific rotation [a]20/D: 7 deg (C=1, CHCl3)
Storage Temperature: 0-10°C
Product Categories: Bioscience, Reagents for Oligosaccharide Synthesis, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-serine tert-Butyl Ester, Fmoc-Ser-OtBu
Formula:
C₂₂H₂₅NO₅ MW: 383.44 g/mol Melting Pt: 127 °C |
MDL Number:
MFCD01861331 CAS Number: 110797-35-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...