4-Amino-p-terphenyl ≥97.0%

Supplier: TCI America

Synonyms: [1,1:4,1-Terphenyl]-4-amine

A1340-5G A1340-1G A1340-25G
TCA1340-5GEA 342.86 CAD
TCA1340-5G TCA1340-001G TCA1340-25G
4-Amino-p-terphenyl ≥97.0%
4-Amino-p-terphenyl

CAS Number: 7293-45-0
MDL Number: MFCD00191302
Molecular Formula: C18H15N
Molecular Weight: 245.33
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 201

Formula: C₁₈H₁₅N
MW: 245.33 g/mol
Melting Pt: 201 °C
MDL Number: MFCD00191302
CAS Number: 7293-45-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2b1c16b79ab6a1fbb05d6b114cbdd3b5c50b5f90&vid=2b1c16b79ab6a1fbb05d6b114cbdd3b5c50b5f90&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR