Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine ≥98.0%

Supplier: TCI America

Synonyms: Fmoc-Asn(Trt)-OH, Nɑ-Fmoc-Nγ-trityl-L-asparagine

F0508-1G F0508-5G F0508-25G
TCF0508-1GEA 33.18 CAD
TCF0508-1G TCF0508-5G TCF0508-25G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-γ-trityl-L-asparagine ≥98.0%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-N-gamma-trityl-L-asparagine

CAS Number: 132388-59-1
Molecular Formula: C38H32N2O5
Molecular Weight: 596.68
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -19 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-Ngamma-trityl-L-asparagine, Fmoc-Asn(Trt)-OH

Formula: C₃₈H₃₂N₂O₅
MDL Number: MFCD00077049
CAS Number: 132388-59-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ff549cea7dcedebdc6fbc0c65dd23bac4dd289ac&vid=ff549cea7dcedebdc6fbc0c65dd23bac4dd289ac&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR