(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98,0

Supplier: TCI America

Synonyms: N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester, Fmoc-Asp(OH)-OtBu, Fmoc-Asp-OtBu, 1-tert-Butyl N-Fmoc-L-aspartate

F0503-1G F0503-5G
TCF0503-1GEA 36.34 CAD
TCF0503-1G TCF0503-5G
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid 98,0
(S)-3-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-4-(tert-butoxy)-4-oxobutanoic acid

CAS Number: 129460-09-9
MDL Number: MFCD00065631
Molecular Formula: C23H25NO6
Molecular Weight: 411.45
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -24 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected), Amino Acids (C-Protected)
Other Product Names / Synonyms: N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic Acid 1-tert-Butyl Ester, 1-tert-Butyl N-Fmoc-L-aspartate, N-Fmoc-L-aspartic Acid 1-tert-Butyl Ester, Fmoc-Asp(OH)-OtBu

Formula: C₂₃H₂₅NO₆
MW: 411.5 g/mol
MDL Number: MFCD00065631
CAS Number: 129460-09-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c860f587d11735249d1543f6184691c0472c1426&vid=c860f587d11735249d1543f6184691c0472c1426&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR