Methyl-6-fluoro-2-naphthoate ≥98.0%

Supplier: TCI America

F0473-1G F0473-5G
TCF0473-1GEA 375.25 CAD
TCF0473-1G TCF0473-5G
Methyl-6-fluoro-2-naphthoate ≥98.0%
Methyl 6-fluoro-2-naphthoate

CAS Number: 5043-00-5
MDL Number: MFCD06797106
Molecular Formula: C12H9FO2
Molecular Weight: 204.20
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 87


Product Categories: Synthetic Organic Chemistry, Fluorinated Building Blocks
Other Product Names / Synonyms: 6-Fluoro-2-naphthoic Acid Methyl Ester

Formula: C₁₂H₉FO₂
Melting Pt: 87 °C
MDL Number: MFCD06797106
CAS Number: 5043-00-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=426514fead14ba98579755dcb97248e8cd6544fa&vid=426514fead14ba98579755dcb97248e8cd6544fa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR