N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine ≥95.0%

Supplier: TCI America

Synonyms: Fmoc-Tyr-OH, N-Fmoc-L-tyrosine

F0456-1G F0456-5G
TCF0456-1GEA 36.34 CAD
TCF0456-1G TCF0456-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine ≥95.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-tyrosine

CAS Number: 92954-90-0
MDL Number: MFCD00134890
Molecular Formula: C24H21NO5
Molecular Weight: 403.43
Purity/Analysis Method: >95.0% (T)
Form: Crystal
Specific rotation [a]20/D: -22 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-tyrosine, Fmoc-Tyr-OH

Formula: C₂₄H₂₁NO₅
MDL Number: MFCD00134890
CAS Number: 92954-90-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4e3350fccbcc743f97627645ae7248c5b11f0572&vid=4e3350fccbcc743f97627645ae7248c5b11f0572&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR