(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid ≥98.0%

Supplier: TCI America

Synonyms: Fmoc-Glu-OH, N-Fmoc-L-glutamic Acid

F0453-1G F0453-5G
TCF0453-1GEA 33.18 CAD
TCF0453-1G TCF0453-5G
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid ≥98.0%
(S)-2-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)pentanedioic acid

CAS Number: 121343-82-6
MDL Number: MFCD00237657
Molecular Formula: C20H19NO6
Molecular Weight: 369.37
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -22 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-glutamic Acid, Fmoc-Glu-OH

Formula: C₂₀H₁₉NO₆
MW: 369.4 g/mol
MDL Number: MFCD00237657
CAS Number: 121343-82-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a049b44aa9e5a80e0b4cb0ebe397ebc851b84235&vid=a049b44aa9e5a80e0b4cb0ebe397ebc851b84235&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR