N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid ≥98.0%
Supplier: TCI America
Synonyms:
N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH
F0452-25G
F0452-5G
F0452-1G
TCF0452-25GEA
224.36
CAD
TCF0452-25G
TCF0452-5G
TCF0452-1G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid ≥98.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-aspartic acid
CAS Number: 119062-05-4
MDL Number: MFCD00237654
Molecular Formula: C19H17NO6
Molecular Weight: 355.35
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -30 deg (C=1, DMF)
Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-L-aspartic Acid, Fmoc-Asp-OH
Formula:
C₁₉H₁₇NO₆ |
MDL Number:
MFCD00237654 CAS Number: 119062-05-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...