4'-Formylbenzo-15-crown-5 (4'-formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0%
Supplier: TCI America
F0448-100MG
F0448-1G
TCF0448-100MGEA
202.24
CAD
TCF0448-100MG
TCF0448-1G
4'-Formylbenzo-15-crown-5 (4'-formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane) ≥98.0%
4'-Formylbenzo-15-crown-5 (4'-Formylbenzo-1,4,7,10,13-pentaoxacyclopentadecane)
CAS Number: 60835-73-6
MDL Number: MFCD00192180
Molecular Formula: C15H20O6
Molecular Weight: 296.32
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 85
Product Categories: Materials Chemistry, Macrocycles for Host-Guest Chemistry
Other Product Names / Synonyms: 2,3-(4-Formylbenzo)-1,4,7,10,13-pentaoxacyclopentadec-2-ene
Formula:
C₁₅H₂₀O₆ Melting Pt: 85 °C |
MDL Number:
MFCD00192180 CAS Number: 60835-73-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...