9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene ≥98.0%

Supplier: TCI America

F0447-25G F0447-5G
TCF0447-25GEA 205.4 CAD
TCF0447-25G TCF0447-5G
9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene ≥98.0%
9,9-Bis[4-(2-hydroxyethoxy)phenyl]fluorene

CAS Number: 117344-32-8
MDL Number: MFCD00216628
Molecular Formula: C29H26O4
Molecular Weight: 438.52
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 163


Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 4,4'-(9-Fluorenylidene)bis(2-phenoxyethanol)

Formula: C₂₉H₂₆O₄
Melting Pt: 163 °C
MDL Number: MFCD00216628
CAS Number: 117344-32-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e47fcc90462da4ff528df188be874a9fc4865869&vid=e47fcc90462da4ff528df188be874a9fc4865869&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR