N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine ≥98.0%

Supplier: TCI America

Synonyms: Fmoc-Phe(4-NO2)-OH, N-Fmoc-4-nitro-L-phenylalanine

F0443-1G F0443-5G
TCF0443-1GEA 48.98 CAD
TCF0443-1G TCF0443-5G
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine ≥98.0%
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-4-nitro-L-phenylalanine

CAS Number: 95753-55-2
MDL Number: MFCD00057810
Molecular Formula: C24H20N2O6
Molecular Weight: 432.43
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -40 deg (C=1, DMF)


Product Categories: Synthetic Organic Chemistry, Peptide Synthesis, Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Fmoc-4-nitro-L-phenylalanine, Fmoc-Phe(4-NO2)-OH

Formula: C₂₄H₂₀N₂O₆
MW: 432.43 g/mol
MDL Number: MFCD00057810
CAS Number: 95753-55-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a37397637ffd8047360fec83c0fdcd73a0772751&vid=a37397637ffd8047360fec83c0fdcd73a0772751&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR