Ethyl (4-fluorobenzoyl)acetate ≥98.0%

Supplier: TCI America

Synonyms: Ethyl 3-(4-Fluorophenyl)-3-oxopropionate, 3-(4-Fluorophenyl)-3-oxopropionic Acid Ethyl Ester, (4-Fluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-fluorophenyl)-3-oxopropanoate

F0435-5G F0435-1G F0435-25G
TCF0435-5GEA 360.24 CAD
TCF0435-5G TCF0435-1G TCF0435-25G
Ethyl (4-fluorobenzoyl)acetate ≥98.0%
Ethyl (4-fluorobenzoyl)acetate

CAS Number: 1999-00-4
MDL Number: MFCD03093631
Molecular Formula: C11H11FO3
Molecular Weight: 210.20
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 120
Flash Point (°C): 110
Specific Gravity (20/20): 1.19


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction), Fluorinated Building Blocks
Other Product Names / Synonyms: (4-Fluorobenzoyl)acetic Acid Ethyl Ester, Ethyl 3-(4-Fluorophenyl)-3-oxopropionate, 3-(4-Fluorophenyl)-3-oxopropionic Acid Ethyl Ester

Formula: C₁₁H₁₁FO₃
MW: 210.20 g/mol
Boiling Pt: 120 °C
Density: 1.19 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD03093631
CAS Number: 1999-00-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ace83f42b4ce5e581a9409ae87312493a98ecc5c&vid=ace83f42b4ce5e581a9409ae87312493a98ecc5c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR