5-Fluoro-2-methylbenzoxazole ≥97.0%

Supplier: TCI America

F0420-5G F0420-1G
TCF0420-5GEA 216.46 CAD
TCF0420-5G TCF0420-001G
5-Fluoro-2-methylbenzoxazole ≥97.0%
5-Fluoro-2-methylbenzoxazole

CAS Number: 701-16-6
MDL Number: MFCD02093471
Molecular Formula: C8H6FNO
Molecular Weight: 151.14
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 90
Specific Gravity (20/20): 1.23

Formula: C₈H₆FNO
Boiling Pt: 90 °C
Density: 1.23 (20 °C)
MDL Number: MFCD02093471
CAS Number: 701-16-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2b71b2383d9e2a5f05d4a50acfeb9c6e37819c4&vid=d2b71b2383d9e2a5f05d4a50acfeb9c6e37819c4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR