4-Formyl-trans-stilbene ≥98.0%

Supplier: TCI America

Synonyms: trans-4-Stilbenecarboxaldehyde

F0412-5G F0412-25G
TCF0412-005GEA 194.34 CAD
TCF0412-005G TCF0412-25G
4-Formyl-trans-stilbene ≥98.0%
4-Formyl-trans-stilbene

CAS Number: 40200-69-9
MDL Number: MFCD00011580
Molecular Formula: C15H12O
Molecular Weight: 208.26
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 114


Other Product Names / Synonyms: trans-4-Stilbenecarboxaldehyde

Formula: C₁₅H₁₂O
MW: 208.26 g/mol
Melting Pt: 114 °C
MDL Number: MFCD00011580
CAS Number: 40200-69-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6589896f6e0cea1d3202380ba5d575127dbe3828&vid=6589896f6e0cea1d3202380ba5d575127dbe3828&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR