2-Fluorophenylboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America

F0407-25G F0407-5G F0407-1G
TCF0407-25GEA 338.12 CAD
TCF0407-25G TCF0407-5G TCF0407-001G
2-Fluorophenylboronic acid (contains varying amounts of Anhydride)
2-Fluorophenylboronic acid

CAS Number: 1993-03-9
MDL Number: MFCD00674013
Molecular Formula: C6H6BFO2
Molecular Weight: 139.92
Form: Crystal
Color: White
Melting point (°C): 110


Other Product Names / Synonyms: 2-Fluorobenzeneboronic Acid

Formula: C₆H₆BFO₂
Melting Pt: 110 °C
MDL Number: MFCD00674013
CAS Number: 1993-03-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=cf2fcd11d3a13907d9848385f75d7ff488e698ef&vid=cf2fcd11d3a13907d9848385f75d7ff488e698ef&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR