Ferroceneacetic acid ≥97.0%
Supplier: TCI America
Synonyms:
Ferrocenylacetic acid
F0406-5G
F0406-1G
TCF0406-5GEA
1442.54
CAD
TCF0406-5G
TCF0406-001G
Ferroceneacetic acid ≥97.0%
Ferroceneacetic acid
CAS Number: 1287-16-7
MDL Number: MFCD00045464
Molecular Formula: C12H12FeO2
Molecular Weight: 244.07
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Formula:
C₁₂H₁₂FeO₂ |
MDL Number:
MFCD00045464 CAS Number: 1287-16-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...