2-(Furfurylthio)ethylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0378-25G F0378-5G
TCF0378-025GEA 270.18 CAD
TCF0378-025G TCF0378-005G
2-(Furfurylthio)ethylamine ≥98.0% (by GC, titration analysis)
2-(Furfurylthio)ethylamine

CAS Number: 36415-21-1
MDL Number: MFCD01075029
Molecular Formula: C7H11NOS
Molecular Weight: 157.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 100
Specific Gravity (20/20): 1.13


Other Product Names / Synonyms: (2-Aminoethylthio)methylfuran

Formula: C₇H₁₁NOS
Boiling Pt: 100 °C
Density: 1.13 (20 °C)
MDL Number: MFCD01075029
CAS Number: 36415-21-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=adaf2777ccc86a11091791addd9f12714fcba9aa&vid=adaf2777ccc86a11091791addd9f12714fcba9aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR