(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine ≥98.0%

Supplier: TCI America

Synonyms: SAMP

A1335-1G A1335-5G
TCA1335-001GEA 267.02 CAD
TCA1335-001G TCA1335-5G
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine ≥98.0%
(S)-(-)-1-Amino-2-(methoxymethyl)pyrrolidine

CAS Number: 59983-39-0
MDL Number: MFCD00064485
Molecular Formula: C6H14N2O
Molecular Weight: 130.19
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 72
Specific Gravity (20/20): 0.98
Specific rotation [a]20/D: -80 deg (neat)


Other Product Names / Synonyms: SAMP

Formula: C₆H₁₄N₂O
MW: 130.19 g/mol
Boiling Pt: 64 °C
Density: 0.98 (20 °C)
Flash Pt: 72 °C
MDL Number: MFCD00064485
CAS Number: 59983-39-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5522fe7b3aa5853df9f18ec893b64f88ec83c157&vid=5522fe7b3aa5853df9f18ec893b64f88ec83c157&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR