Cyanomethyl formate ≥98.0%
Supplier: TCI America
F0356-25G
F0356-5G
TCF0356-25GEA
813.7
CAD
TCF0356-25G
TCF0356-005G
Cyanomethyl formate ≥98.0%
Cyanomethyl formate
[Formylating Reagent]
CAS Number: 150760-95-5
MDL Number: MFCD00661077
Molecular Formula: C3H3NO2
Molecular Weight: 85.06
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 64
Flash Point (°C): 82
Specific Gravity (20/20): 1.19
Product Categories: Synthetic Organic Chemistry, Formylation
Other Product Names / Synonyms: Formic Acid Cyanomethyl Ester
Formula:
C₃H₃NO₂ Boiling Pt: 64 °C Density: 1.19 (20 °C) Flash Pt: 82 °C |
MDL Number:
MFCD00661077 CAS Number: 150760-95-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...