Methyl fumaraldehydate ≥95.0%

Supplier: TCI America

F0351-100MG F0351-1G
TCF0351-100MGEA 101.12 CAD
TCF0351-100MG TCF0351-1G
Methyl fumaraldehydate ≥95.0%
Methyl fumaraldehydate

CAS Number: 5837-72-9
MDL Number: MFCD00191651
Molecular Formula: C5H6O3
Molecular Weight: 114.10
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Boiling point (°C): 88
Melting point (°C): 40
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 3-(Methoxycarbonyl)-2-propenal, Fumaraldehydic Acid Methyl Ester, 3-(Methoxycarbonyl)acrylaldehyde

Formula: C₅H₆O₃
Boiling Pt: 88 °C
Melting Pt: 40 °C
MDL Number: MFCD00191651
CAS Number: 5837-72-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fa29b490c4d44d78074cb6a691030a15e2a21188&vid=fa29b490c4d44d78074cb6a691030a15e2a21188&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR