Fumaraldehyde mono(dimethyl acetal) ≥95.0% stabilised

Supplier: TCI America

Synonyms: 4,4-Dimethoxycrotonaldehyde, 4,4-Dimethoxy-2-butenal

F0350-1G F0350-5G
TCF0350-001GEA 289.3 CAD
TCF0350-001G TCF0350-5G
Fumaraldehyde mono(dimethyl acetal) ≥95.0% stabilised
Fumaraldehyde mono(dimethyl acetal)

(stabilized with HQ)
CAS Number: 4093-49-6
MDL Number: MFCD00191439
Molecular Formula: C6H10O3
Molecular Weight: 130.14
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Specific Gravity (20/20): 1.03
Storage Temperature: 0-10°C


Other Product Names / Synonyms: 4,4-Dimethoxy-2-butenal, 4,4-Dimethoxycrotonaldehyde

Formula: C₆H₁₀O₃
Density: 1.03 (20 °C)
MDL Number: MFCD00191439
CAS Number: 4093-49-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5df938005f88f20c0468f18c519bfe26783139ec&vid=5df938005f88f20c0468f18c519bfe26783139ec&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR