Fluoromethyl phenyl sulfone ≥98.0%

Supplier: TCI America

Synonyms: ((Fluoromethyl)sulfonyl)benzene, Fluoromethylsulfonylbenzene, Fluoromethylsulphonylbenzene, ((Fluoromethyl)sulphonyl)benzene

F0341-5G F0341-1G
TCF0341-5GEA 1098.1 CAD
TCF0341-5G TCF0341-001G
Fluoromethyl phenyl sulfone ≥98.0%
Fluoromethyl phenyl sulfone

CAS Number: 20808-12-2
MDL Number: MFCD00191650
Molecular Formula: C7H7FO2S
Molecular Weight: 174.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 53


Product Categories: Synthetic Organic Chemistry, Sulfones, Bioscience, Reagents for Oligosaccharide Synthesis
Other Product Names / Synonyms: Fluoromethylsulfonylbenzene

Formula: C₇H₇FO₂S
MW: 174.19 g/mol
Melting Pt: 53 °C
MDL Number: MFCD00191650
CAS Number: 20808-12-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f407cbc0bac1691ca0466ef1c2845c08193e021&vid=4f407cbc0bac1691ca0466ef1c2845c08193e021&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR