5-Formyl-2-furancarboxylic acid ≥98.0%
Supplier: TCI America
Synonyms:
Furfural-5-carboxylic Acid, 5-Formylfuran-2-carboxylic acid, 5-Formyl-2-furoic Acid
F0338-1G
F0338-5G
TCF0338-001GEA
287.56
CAD
TCF0338-001G
TCF0338-005G
5-Formyl-2-furancarboxylic acid ≥98.0%
5-Formyl-2-furancarboxylic acid
CAS Number: 13529-17-4
MDL Number: MFCD00020924
Molecular Formula: C6H4O4
Molecular Weight: 140.09
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 209
Other Product Names / Synonyms: 5-Formyl-2-furoic Acid, Furfural-5-carboxylic Acid
Formula:
C₆H₄O₄ MW: 140.09 g/mol Melting Pt: 209 °C |
MDL Number:
MFCD00020924 CAS Number: 13529-17-4 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...