3-Fluoro-4-n-octyloxybenzoic acid ≥97.0%

Supplier: TCI America

F0337-5G F0337-1G
TCF0337-5GEA 1376.81 CAD
TCF0337-5G TCF0337-001G
3-Fluoro-4-n-octyloxybenzoic acid ≥97.0%
3-Fluoro-4-n-octyloxybenzoic acid

CAS Number: 326-78-3
MDL Number: MFCD00191649
Molecular Formula: C15H21FO3
Molecular Weight: 268.33
Purity/Analysis Method: >97.0% (GC,T)
Form: Crystal

Formula: C₁₅H₂₁FO₃
MW: 268.33 g/mol
MDL Number: MFCD00191649
CAS Number: 326-78-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60eecbf96e8cf87e4329eb3f9e8344726e6e575&vid=f60eecbf96e8cf87e4329eb3f9e8344726e6e575&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR