5-Formyl-2-thiophenecarboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

F0336-1G F0336-5G
TCF0336-001GEA 135.88 CAD
TCF0336-001G TCF0336-005G
5-Formyl-2-thiophenecarboxylic acid ≥98.0% (by GC, titration analysis)
5-Formyl-2-thiophenecarboxylic acid

CAS Number: 4565-31-5
MDL Number: MFCD00191437
Molecular Formula: C6H4O3S
Molecular Weight: 156.16
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 169


Other Product Names / Synonyms: 5-Formylthenoic Acid

Formula: C₆H₄O₃S
Melting Pt: 169 °C
MDL Number: MFCD00191437
CAS Number: 4565-31-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c3daa8574031ddf1dac618546aae6d15d32708fb&vid=c3daa8574031ddf1dac618546aae6d15d32708fb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR