Fumaraldehydebis(dimethyl acetal) ≥90.0%
Supplier: TCI America
Synonyms:
Fumaraldehyde bis(dimethylacetal), 1,1,4,4-Tetramethoxy-trans-2-butene, (E)-1,1,4,4-Tetramethoxybut-2-ene
F0329-1G
F0329-5G
TCF0329-001GEA
62.88
CAD
TCF0329-001G
TCF0329-5G
Fumaraldehydebis(dimethyl acetal) ≥90.0%
Fumaraldehydebis(dimethyl acetal)
CAS Number: 6068-62-8
MDL Number: MFCD00075507
Molecular Formula: C8H16O4
Molecular Weight: 176.21
Purity/Analysis Method: >90.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 80
Specific Gravity (20/20): 1.01
Other Product Names / Synonyms: 1,1,4,4-Tetramethoxy-trans-2-butene
Formula:
C₈H₁₆O₄ MW: 176.21 g/mol Boiling Pt: 103 °C Density: 1.01 (20 °C) Flash Pt: 80 °C |
MDL Number:
MFCD00075507 CAS Number: 6068-62-8 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...