Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine ≥98.0%

Supplier: TCI America

Synonyms: Nα-Fmoc-L-glutamine, Nɑ-Fmoc-L-glutamine, Fmoc-Gln-OH

F0308-1G F0308-5G
TCF0308-001GEA 31.6 CAD
TCF0308-001G TCF0308-005G
Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine ≥98.0%
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-L-glutamine

CAS Number: 71989-20-3
MDL Number: MFCD00037137
Molecular Formula: C20H20N2O5
Molecular Weight: 368.39
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Specific rotation [a]20/D: -18 deg (C=1, DMF)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: Nalpha-Fmoc-L-glutamine, Fmoc-Gln-OH

Formula: C₂₀H₂₀N₂O₅
MW: 368.39 g/mol
MDL Number: MFCD00037137
CAS Number: 71989-20-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3de96586ce99f042165a735bffcd2eacfedb7e9a&vid=3de96586ce99f042165a735bffcd2eacfedb7e9a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR